missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Benzoic sulfimide sodium salt hydrate, 99%

Manufacturer:  Alfa Aesar™ A155300I

 View more versions of this product

Catalog No. AAA155300I



Description

Description

Applications
O-benzoic sulfimide sodium salt is used as an artificial sweetener in drinks, candies, cookies, medicines and toothpaste. It is used in the food industry and for sweetening diabetes diet. It is also used as a synthetic sweet ignorance agent. It finds application in a variety of beverages, foods, cosmetics and pharmaceuticals to provide sweetness without adding calories. It is involved as a nickel electroplating brightener, chemical intermediate, animal feed sweetener and anaerobic adhesive accelerator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

o-Benzoic sulfimide sodium salt hydrate
5000g
C7H9NNaO4S
3599229
saccharin sodium salt hydrate
[HH].C1=CC=C2C(=C1)C(=O)NS2(=O)=O.O.[Na]
226.202
205.16 (Anhydrous)
>300°C
82385-42-0
MFCD00151213
14,8312
NPCHGKCOTXZNMS-UHFFFAOYSA-N
1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate
131673955
99%

Chemical Identifiers

82385-42-0
226.202
NPCHGKCOTXZNMS-UHFFFAOYSA-N
131673955
[HH].C1=CC=C2C(=C1)C(=O)NS2(=O)=O.O.[Na]
C7H9NNaO4S
MFCD00151213
saccharin sodium salt hydrate
1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate
Safety and Handling

Safety and Handling

RTECSNumber : DE4550000

TSCA : TSCA

Documents