Learn More
o-Benzoic sulfimide, 98+%, Thermo Scientific Chemicals
$55.27 - $535.29
Chemical Identifiers
CAS | 81-07-2 |
---|---|
Molecular Formula | C7H5NO3S |
Molecular Weight (g/mol) | 183.181 |
MDL Number | MFCD00005866 |
InChI Key | CVHZOJJKTDOEJC-UHFFFAOYSA-N |
Synonym | saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose |
PubChem CID | 5143 |
ChEBI | CHEBI:32111 |
IUPAC Name | 1,1-dioxo-1,2-benzothiazol-3-one |
SMILES | C1=CC=C2C(=C1)C(=O)NS2(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2393822
|
Thermo Scientific Chemicals
B2393822 |
100 g |
N/A
|
|
|||||
AAB2393836
|
Thermo Scientific Chemicals
B2393836 |
500 g |
N/A
|
|
|||||
AAB239380E
|
Thermo Scientific Chemicals
B239380E |
2500 g |
N/A
|
|
|||||
Description
o-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationso-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.
Solubility
Soluble in water (3.3 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
81-07-2 | |
183.181 | |
CVHZOJJKTDOEJC-UHFFFAOYSA-N | |
5143 | |
1,1-dioxo-1,2-benzothiazol-3-one |
C7H5NO3S | |
MFCD00005866 | |
saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose | |
CHEBI:32111 | |
C1=CC=C2C(=C1)C(=O)NS2(=O)=O |
Specifications
81-07-2 | |
0.828 | |
MFCD00005866 | |
6888 | |
saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose | |
CVHZOJJKTDOEJC-UHFFFAOYSA-N | |
1,1-dioxo-1,2-benzothiazol-3-one | |
5143 | |
183.18 | |
o-Benzoic sulfimide |
227°C to 230°C | |
C7H5NO3S | |
100 g | |
14,8311 | |
Soluble in water (3.3g/L at 20°C). | |
C1=CC=C2C(=C1)C(=O)NS2(=O)=O | |
183.181 | |
CHEBI:32111 | |
≥98% |
Safety and Handling
EINECSNumber : 201-321-0
RTECSNumber : DE4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only