missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Anisidine 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 104812500

 View more versions of this product

Catalog No. AC104812500


 

Specifications Description & Specifications

Specifications

≥99.0%
123.15
Clear colorless to light yellow-brown
14, 667
100.0
2-Methoxyaniline
CH3OC6H4NH2
C7H9NO
o-Anisidine
Glass bottle
This substance darkens on exposure to air
Liquid
250mL
13, 358
99.0
1.5730 to 1.5750 (20°C, 589nm)
99+%
99+%
MFCD00007688
90-04-0
Authentic

Chemical Identifiers

90-04-0
100.0
C7H9NO
123.15
2-Methoxyaniline
≥99.0%
99.0
99+%
CH3OC6H4NH2
MFCD00007688
o-Anisidine, 99+%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause cancer.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-963-1

RTECSNumber : BZ5410000

TSCA : TSCA