missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Anisidine, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A102880I

 View more versions of this product

Catalog No. AAA102880I



Description

Description

Applications
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
Specifications

Specifications

o-Anisidine
99%
C7H9NO
UN2431
14,667
VMPITZXILSNTON-UHFFFAOYSA-N
2-methoxyaniline
7000
123.16
5°C to 6°C
98°C (208°F)
90-04-0
99%
MFCD00007688
386210
2-Aminoanisole; 2-Methoxyaniline
COC1=CC=CC=C1N
123.155
CHEBI:82288
224°C to 225°C
5000g

Chemical Identifiers

90-04-0
123.155
VMPITZXILSNTON-UHFFFAOYSA-N
7000
2-methoxyaniline
C7H9NO
MFCD00007688
2-Aminoanisole; 2-Methoxyaniline
CHEBI:82288
COC1=CC=CC=C1N
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H350-H341
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Suspected of causing genetic defects.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANISIDINES

RTECSNumber : BZ5410000

TSCA : TSCA

SDS Documents