Learn More
o-Anisaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 149221000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
o-Anisaldehyde | |
135-02-4 | |
100.0 | |
Brown to Yellow | |
117°C | |
97.5% min. (GC) | |
C8H8O2 | |
CH3OC6H4CHO | |
100 g | |
10, 5866 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol | |
COC1=CC=CC=C1C=O | |
136.15 | |
136.15 | |
Crystalline Low Melting Solid |
98% | |
97.5 | |
34.0°C to 40.0°C | |
238.0°C | |
Authentic | |
Glass bottle | |
1.5608 | |
MFCD00003308 | |
08, IV, 180 | |
o-anisaldehyde, benzaldehyde, 2-methoxy, 2-anisaldehyde, o-methoxybenzaldehyde, 6-methoxybenzaldehyde, salicylaldehyde methyl ether, 2-methoxybenzenecarboxaldehyde, benzaldehyde, o-methoxy, 2-methoxyphenylformaldehyde, formylanisole, o | |
PKZJLOCLABXVMC-UHFFFAOYSA-N | |
2-methoxybenzaldehyde | |
8658 | |
98% |
Chemical Identifiers
135-02-4 | |
136.15 | |
PKZJLOCLABXVMC-UHFFFAOYSA-N | |
8658 | |
COC1=CC=CC=C1C=O |
C8H8O2 | |
MFCD00003308 | |
o-anisaldehyde, benzaldehyde, 2-methoxy, 2-anisaldehyde, o-methoxybenzaldehyde, 6-methoxybenzaldehyde, salicylaldehyde methyl ether, 2-methoxybenzenecarboxaldehyde, benzaldehyde, o-methoxy, 2-methoxyphenylformaldehyde, formylanisole, o | |
2-methoxybenzaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 205-171-7
RTECSNumber : BZ2610000
TSCA : TSCA
RUO – Research Use Only