missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Acetylsalicyloyl chloride, 97%

Manufacturer:  Alfa Aesar™ L0091922

 View more versions of this product

Catalog No. AAL0091922



Description

Description

Applications
O-Acetylsalicyloyl chloride was used in the synthesis of enantiomerically pure bidentate heteroorganic ligands built on simple achiral skeletons and containing an aziridine moiety. It was used in the general synthesis of acetoxybenzamides. It was used in the chemical synthesis of 2,2,6,6-tetramethyl-1-piperidinyloxy(TEMPO)-aspirin conjugate via condensation reaction with 4-hydroxy-TEMPO. It was used as reagent in acylation of cyclobutenediones.

Solubility
Soluble in toluene. Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from strong bases.
Specifications

Specifications

O-Acetylsalicyloyl chloride
Pungent, Makes Eyes Water
134°C to 136°C (12mmHg)
100g
C9H7ClO3
UN3261
2-acetoxybenzoyl chloride, 2-carbonochloridoyl phenyl acetate, 2-chlorocarbonyl phenyl acetate, acetylsalicoyl chloride, acetylsalicyloyl chloride, ascc, aspirin chloride, benzoyl chloride, 2-acetyloxy, o-acetylsalicyloyl chloride, o-acetylsalicyloylchloride
DSGKWFGEUBCEIE-UHFFFAOYSA-N
(2-carbonochloridoylphenyl) acetate
79668
97%
>110°C (230°F)
Moisture sensitive
47°C to 50°C
5538-51-2
MFCD00000663
880372
Soluble in toluene. Reacts with water.
CC(=O)OC1=CC=CC=C1C(=O)Cl
198.602
198.61

Chemical Identifiers

5538-51-2
198.602
DSGKWFGEUBCEIE-UHFFFAOYSA-N
79668
CC(=O)OC1=CC=CC=C1C(=O)Cl
C9H7ClO3
MFCD00000663
2-acetoxybenzoyl chloride, 2-carbonochloridoyl phenyl acetate, 2-chlorocarbonyl phenyl acetate, acetylsalicoyl chloride, acetylsalicyloyl chloride, ascc, aspirin chloride, benzoyl chloride, 2-acetyloxy, o-acetylsalicyloyl chloride, o-acetylsalicyloylchloride
(2-carbonochloridoylphenyl) acetate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 226-899-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents