missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Acetyl-L-carnitine hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 342390010

 View more versions of this product

Catalog No. AC342390010


 

Specifications Description & Specifications

Specifications

≥97.5%
239.7
White
100.0
98%
98%
MFCD00082230
o-Acetyl-L-carnitine hydrochloride
-27° to -31° (20°C, 589nm) (c=1, H2O)
Crystalline Powder
1g
15, 79
Authentic
5080-50-2
C9H17NO4·HCl
97.5
Glass bottle
Levacecarnine hydrochloride

Chemical Identifiers

5080-50-2
100.0
C9H17NO4·HCl
MFCD00082230
o-Acetyl-L-carnitine hydrochloride, 98%
97.5
98%
239.7
Levacecarnine hydrochloride
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC