Please login to your online account to display your discounted pricing

o-Acetyl-L-carnitine hydrochloride, 98%, ACROS Organics™

$39.92 - $39.92

Chemical Identifiers

Chemical Name or Material o-Acetyl-L-carnitine hydrochloride
Name Note 98%
CAS 5080-50-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H17NO4·HCl
Formula Weight 239.7
MDL Number MFCD00082230
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC342390010

  SDS
thermo fisher scientific
342390010
Glass bottle 1g Each for $39.92
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 239.7
Quantity 1g
Color White
Merck Index 15, 79
CAS Max % 100.0
Infrared Spectrum Authentic
Name Note 98%
CAS 5080-50-2
Assay Percent Range 98%
Molecular Formula C9H17NO4·HCl
MDL Number MFCD00082230
CAS Min % 97.5
Chemical Name or Material o-Acetyl-L-carnitine hydrochloride
Packaging Glass bottle
Specific Rotation -27° to -31° (20°C, 589nm) (c=1, H2O)
Synonym Levacecarnine hydrochloride
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 1 HYGROSCOPIC
Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.