missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
o-Acetotoluidine 98.0+%, TCI America™
$69.68 - $69.68
Chemical Identifiers
CAS | 120-66-1 |
---|---|
Molecular Formula | C9H11NO |
Molecular Weight (g/mol) | 149.193 |
MDL Number | MFCD00014961 |
InChI Key | BPEXTIMJLDWDTL-UHFFFAOYSA-N |
Synonym | o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide |
PubChem CID | 8443 |
IUPAC Name | N-(2-methylphenyl)acetamide |
SMILES | CC1=CC=CC=C1NC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A006325G
|
TCI America
A006325G |
25 g |
Each for $69.68
|
|
|||||
Chemical Identifiers
120-66-1 | |
149.193 | |
BPEXTIMJLDWDTL-UHFFFAOYSA-N | |
8443 | |
CC1=CC=CC=C1NC(=O)C |
C9H11NO | |
MFCD00014961 | |
o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide | |
N-(2-methylphenyl)acetamide |
Specifications
120-66-1 | |
Gray-White | |
C9H11NO | |
25 g | |
BPEXTIMJLDWDTL-UHFFFAOYSA-N | |
N-(2-methylphenyl)acetamide | |
8443 | |
≥98.0% (GC) | |
o-Acetotoluidine |
110°C | |
296°C | |
MFCD00014961 | |
o-acetotoluidide, 2'-methylacetanilide, n-o-tolyl acetamide, n-2-methylphenyl acetamide, acetyl-o-toluidine, n-acetyl-o-toluidine, o-acetotoluide, acetamide, n-2-methylphenyl, o-methylacetanilide, n-o-tolylacetamide | |
CC1=CC=CC=C1NC(=O)C | |
149.193 | |
149.19 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : AN2900000
TSCA : Yes