Learn More
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2608203
Description
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-(7-Aza-1H-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.
Solubility
Soluble in acetonitrile. Insoluble in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents and highly acidic or alkaline materials.
Specifications
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate | |
∼185°C (decomposition) | |
MFCD00274639 | |
Moisture sensitive | |
Soluble in acetonitrile. Insoluble in water. | |
F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C | |
380.24 | |
380.23 |
148893-10-1 | |
C10H15F6N6OP | |
1 g | |
hatu, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n,n-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, dimethylamino triazolo 4,5-b pyridin-3-yloxy methylene-dimethyl-ammonium hexafluorophosphate, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, n-dimethylamino 3h-1,2,3 triazolo 4,5-b pyridin-3-yloxy methylene-n-methylmethanaminium hexafluorophosphate, n-dimethylamino-1h-1,2,3-triazolo-4,5-b pyridin-1-ylmethylene-n-methylmethanaminium hexafluorophosphate n-oxide | |
JNWBBCNCSMBKNE-UHFFFAOYSA-N | |
[dimethylamino(triazolo[4,5-b]pyridin-3-yloxy)methylidene]-dimethylazanium;hexafluorophosphate | |
9886157 | |
99% |
Chemical Identifiers
148893-10-1 | |
380.24 | |
JNWBBCNCSMBKNE-UHFFFAOYSA-N | |
9886157 | |
F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C |
C10H15F6N6OP | |
MFCD00274639 | |
hatu, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n,n-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, dimethylamino triazolo 4,5-b pyridin-3-yloxy methylene-dimethyl-ammonium hexafluorophosphate, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, n-dimethylamino 3h-1,2,3 triazolo 4,5-b pyridin-3-yloxy methylene-n-methylmethanaminium hexafluorophosphate, n-dimethylamino-1h-1,2,3-triazolo-4,5-b pyridin-1-ylmethylene-n-methylmethanaminium hexafluorophosphate n-oxide | |
[dimethylamino(triazolo[4,5-b]pyridin-3-yloxy)methylidene]-dimethylazanium;hexafluorophosphate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Hazard Class: 4.1; Packaging Group: II
RTECSNumber : XZ5633000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only