missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 99+%

Manufacturer:  Alfa Aesar™ H5473222

 View more versions of this product

Catalog No. AAH5473222



Specifications

Specifications

O-(6-Chloro-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate
100g
C11H15BClF4N5O
1h-benzotriazolium, 1-bis dimethylamino methylene-5-chloro-, 3-oxide tetrafluoroborate 1-1:1
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C2=C(C=CC(=C2)Cl)N=N1
355.529
355.53
198°C to 204°C
330641-16-2
MFCD04973270
GBGVQFJZGHBZMC-UHFFFAOYSA-N
[(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;tetrafluoroborate
24898642
≥99%

Chemical Identifiers

330641-16-2
355.529
GBGVQFJZGHBZMC-UHFFFAOYSA-N
24898642
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C2=C(C=CC(=C2)Cl)N=N1
C11H15BClF4N5O
MFCD04973270
1h-benzotriazolium, 1-bis dimethylamino methylene-5-chloro-, 3-oxide tetrafluoroborate 1-1:1
[(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

Documents