missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H6331206

 View more versions of this product

Catalog No. AAH6331206



Description

Description

Applications
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

O-(4-Methoxybenzyl)hydroxylamine hydrochloride
5g
C8H11NO2
o-4-methoxybenzyl hydroxylamine, o-4-methoxyphenyl methyl hydroxylamine, p-methoxybenzyloxyamine, hydroxylamine, o-p-methoxybenzyl, hydroxylamine, o-4-methoxyphenyl methyl, o-p-methoxybenzyl hydroxylamine, o-p-methoxybenzyl-hydroxylamine, o-4-methoxy-benzyl-hydroxylamine, 1-aminooxy methyl-4-methoxybenzene, 1-aminooxy methyl 4-methoxy-benzene
COC1=CC=C(C=C1)CON
153.181
153.18
98%
White
21038-22-2
MFCD01114582
MVSMBIBGGPSEHQ-UHFFFAOYSA-N
O-[(4-methoxyphenyl)methyl]hydroxylamine
120426
Solid

Chemical Identifiers

21038-22-2
153.181
MVSMBIBGGPSEHQ-UHFFFAOYSA-N
120426
COC1=CC=C(C=C1)CON
C8H11NO2
MFCD01114582
o-4-methoxybenzyl hydroxylamine, o-4-methoxyphenyl methyl hydroxylamine, p-methoxybenzyloxyamine, hydroxylamine, o-p-methoxybenzyl, hydroxylamine, o-4-methoxyphenyl methyl, o-p-methoxybenzyl hydroxylamine, o-p-methoxybenzyl-hydroxylamine, o-4-methoxy-benzyl-hydroxylamine, 1-aminooxy methyl-4-methoxybenzene, 1-aminooxy methyl 4-methoxy-benzene
O-[(4-methoxyphenyl)methyl]hydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NC3676000

SDS Documents