Please login to your online account to display your discounted pricing

Alfa Aesar™ O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 99+%

$197.39 - $197.39

Chemical Identifiers

Chemical Name or Material O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride
CAS 57981-02-9
Molecular Formula C7H4F5NO·HCl
Formula Weight 249.57
MDL Number MFCD00012953
Beilstein 4031190
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1836803

  SDS
alfa aesar™
A1836803
1g Each for $197.39

AAA1836801

  SDS
alfa aesar™
A1836801
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1836806

  SDS
alfa aesar™
A1836806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride
CAS 57981-02-9
Assay ≥99%
Molecular Formula C7H4F5NO·HCl
Formula Weight 249.57
MDL Number MFCD00012953
Beilstein 4031190
Melting Point ≈215°C (subl.)
Quantity 1g

Applications
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride is used in the preparation of oximes of steroids having keto group. It is used as a derivatization reagent in the determination of thromboxane B2, prostaglandins, amygdalin and a variety of aldehydes, ketones and acids. It is a versatile reagent utilized for the detection of carbonyl-containing compounds by gas chromatography (GC), gas chromatography mass spectrometry (GC-MS) and other methods. As a derivatization reagent, it is also used for EC/LC analysis of keto-steroids and carbohydrates.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.