Please login to your online account to display your discounted pricing

Alfa Aesar™ O(1),O(3)-Bis(ethoxycarbonylmethyl)-p-tert-butylcalix[4]arene

$100.81 - $405.32

Chemical Identifiers

Chemical Name or Material O(1),O(3)-Bis(ethoxycarbonylmethyl)-p-tert-butylcalix[4]arene
CAS 97600-49-2
Molecular Formula C52H68O8
Formula Weight 821.11
MDL Number MFCD00798575
Synonym O(1),O(3)-Bis(carbethoxymethyl)-p-tert-butylcalix[4]arene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1959402

  SDS
alfa aesar™
A1959402
0.5g Each for $405.32

AAA1959477

  SDS
alfa aesar™
A1959477
0.1g Each for $100.81
Description & Specifications

Specifications

Formula Weight 821.11
Chemical Name or Material O(1),O(3)-Bis(ethoxycarbonylmethyl)-p-tert-butylcalix[4]arene
Molecular Formula C52H68O8
MDL Number MFCD00798575
CAS 97600-49-2
Synonym O(1),O(3)-Bis(carbethoxymethyl)-p-tert-butylcalix[4]arene
Melting Point 182° to 184°C
Quantity 0.5g

Applications
O(1),O(3)-Bis(ethoxycarbonylmethyl)-p-tert-butylcalix[4]arene is used as an intermediates, amino acid and other fine chemicals.

Solubility
Sparingly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.