Please login to your online account to display your discounted pricing

Alfa Aesar™ O(1),O(3)-Bis(ethoxycarbonylmethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene

$102.99 - $414.07

Chemical Identifiers

Chemical Name or Material O(1),O(3)-Bis(ethoxycarbonylmethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene
CAS 149775-71-3
Molecular Formula C54H72O8
Formula Weight 849.16
MDL Number MFCD00798577
Synonym O(1),O(3)-Bis(carbethoxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1968802

 
alfa aesar™
A1968802
0.5g Each for $414.07

AAA1968877

 
alfa aesar™
A1968877
0.1g Each for $102.99
Description & Specifications

Specifications

Formula Weight 849.16
Chemical Name or Material O(1),O(3)-Bis(ethoxycarbonylmethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene
Molecular Formula C54H72O8
MDL Number MFCD00798577
CAS 149775-71-3
Synonym O(1),O(3)-Bis(carbethoxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene
Melting Point 162° to 164°C
Quantity 0.5g