missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O(1),O(3)-Bis(carboxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1919277

 View more versions of this product

Catalog No. AAA1919277



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

O(1),O(3)-Bis(carboxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene
250°C to 252°C
MFCD00798565
o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene, o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-t-butylcalix 4 arene
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3OCC(=O)O)CC4=CC(=CC(=C4OC)CC5=C(C(=CC(=C5)C(C)(C)C)C2)OCC(=O)O)C(C)(C)C)C(C)(C)C)OC
15243456
136157-98-7
C50H64O8
0.1 g
GRSQCGICFHAMOA-UHFFFAOYSA-N
793.054
793.06

Chemical Identifiers

136157-98-7
793.054
GRSQCGICFHAMOA-UHFFFAOYSA-N
15243456
C50H64O8
MFCD00798565
o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene, o 1 ,o 3-bis carboxymethyl-o 2 ,o 4-dimethyl-p-t-butylcalix 4 arene
CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3OCC(=O)O)CC4=CC(=CC(=C4OC)CC5=C(C(=CC(=C5)C(C)(C)C)C2)OCC(=O)O)C(C)(C)C)C(C)(C)C)OC
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only