Please login to your online account to display your discounted pricing

Tocris™ NXY 059

Free radical trapping agent; neuroprotectant

$164.61 - $682.67

Specifications

Product Type NXY 059
Inhibitors NXY 059
Molecular Weight 381.33
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

364510R

 
tocris™
RDS364510
10mg Each for $164.61

364550R

 
tocris™
RDS364550
50mg Each for $682.67
Description & Specifications

Specifications

Product Type NXY 059
Inhibitors NXY 059
Molecular Weight 381.33
Purity >98

Free radical trapping agent. Reduces infarct size and preserves brain function in several animal models of acute ischemic stroke. Neuroprotectant.

  • Chemical Name: 4-[[(1,1-Dimethylethyl)oxoimino]methyl]-1,3-benzenedisulfonic acid disodium salt
  • Formula: C11H13NNa2O7S2
  • CAS Number: 168021-79-2
  • Solubility: Soluble to 100mM in water and to 100mM in DMSO
  • Purity: >98