missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Newtex™ NXP 150 Approach Suit

$588.45 - $3,218.57

Specifications

Certifications/Compliance ISO 6942, EN 366, ISO 9151, EN 367, ISO 15025, EN 532, ISO 9185, EN 348 and ASTM F955, ISO 13937, EN 388, NFPA 701, ASTM D6413
Color Silver
Material Z-Flex™ Aluminized Fabric
Liner Material Cotton
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-986-13C
View Documents Promotion Details
Newtex™
2100099
X-Large
Each for $588.45
 
17-988-404A
View Documents Promotion Details
Newtex™
2100051
Medium
Each for $3,218.57
 
17-988-404B
View Documents Promotion Details
Newtex™
2100050
Large
Each for $3,188.22
 
Description

Description

  • Made from Z-Flex multilayer aluminized fabric
  • Provides heat protection where ambient temperature does not exceed 200°F (93°C )
  • Aluminized coating reflects 90% of radiant heat, so garment is suitable for intermittent exposure to radiant heat up to 2000°F (1093°C)
  • Flame-retardant cotton lining adds protection and comfort
  • Large-size standard suit weighs 16 lb. (7.3kg)
  • tandard suit contains five components: hood, coat, pants, boots, and gloves

Hood. A hard cap attached to built-in fiberglass support. “Speedy clip” attachment allows easy removal. Hood has shoulder drape, underarm straps, and 6 × 9 1/4 in. (15.2 × 23.5cm) window of gold film over Plexiglas™ plastic. Window has aluminum retainer and two wing locks.Coat. Length, 32 in. (81cm). Tailored sleeves with snap adjustment. Covered snap closure.Pants. High waist, eight-point elastic suspenders, and snapped triangular storm-fly front.Boots. Fit over work shoes and have leather soles and heels. Adjustable tightening straps.Gloves. Length, 14 in. (36cm). Gauntlet style with leather palms. SEF™ fire-retardant lining in fingers.

Note:As special order, suit is available with expanded back, which accommodates SCBA; hood, gloves, and boots also available separately.

Order Info

Coat and pants available separately. Ask your Fisher Safety Customer Service Representative for details.

Specifications

Specifications

ISO 6942, EN 366, ISO 9151, EN 367, ISO 15025, EN 532, ISO 9185, EN 348 and ASTM F955, ISO 13937, EN 388, NFPA 701, ASTM D6413
Silver
Z-Flex™ Aluminized Fabric
Cotton
SDS
Product Certifications