Please login to your online account to display your discounted pricing

NVP DPP 728 dihydrochloride, Tocris™

Potent, orally active dipeptidyl peptidase IV (DPP-IV) inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material NVP DPP 728 dihydrochloride
Assay Percent Range >98%
Molecular Formula C15H18N6O·2HCl
Formula Weight 371.27
Synonym 6-[[2-[[2-(2S)-2-Cyano-1-pyrrolidinyl]-2-oxoethyl]amino]ethyl]amino-3-pyridinecarbononitrile dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

350610

 
r & d systems
RDS350610
10mg Each for $252.39

350650

 
r & d systems
RDS350650
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 371.27
Quantity 10mg
CAS 207556-62-5
Molecular Formula C15H18N6O·2HCl
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material NVP DPP 728 dihydrochloride
Assay Percent Range >98%
Synonym 6-[[2-[[2-(2S)-2-Cyano-1-pyrrolidinyl]-2-oxoethyl]amino]ethyl]amino-3-pyridinecarbononitrile dihydrochloride
Safety and Handling

Desiccate at Room Temperature