Please login to your online account to display your discounted pricing

NVP 231, Tocris™

Potent, selective and reversible CerK inhibitor

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material NVP 231
Assay Percent Range >98%
Molecular Formula C25H25N3O2S
Formula Weight 431.55
Synonym N-[2-(Benzoylamino)-6-benzothiazolyl]tricyclo[3.3.1.13, 7]decane-1-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

396010

 
r & d systems
RDS396010
10mg Each for $218.74

396050

 
r & d systems
RDS396050
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 431.55
Quantity 10mg
Chemical Name or Material NVP 231
Assay Percent Range >98%
Molecular Formula C25H25N3O2S
CAS 362003-83-6
Synonym N-[2-(Benzoylamino)-6-benzothiazolyl]tricyclo[3.3.1.13, 7]decane-1-carboxamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C