Please login to your online account to display your discounted pricing

Nutlin-3, Tocris™

MDM2 antagonist; inhibits MDM2-p53 interaction

$316.20 - $1,307.92

Chemical Identifiers

Chemical Name or Material Nutlin-3
Assay Percent Range >98%
Molecular Formula C30H30Cl2N4O4
Formula Weight 581.49
Synonym (±)-4-[4, 5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxy-phenyl)-4, 5-dihydro-imidazole-1-carbonyl]-piperazin-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

398410

 
r & d systems
398410
10mg Each for $316.20

398450

 
r & d systems
398450
50mg Each for $1,307.92
Description & Specifications

Specifications

Formula Weight 581.49
Quantity 10mg
Chemical Name or Material Nutlin-3
CAS 548472-68-0
Assay Percent Range >98%
Recommended Storage Store at -20°C
Synonym (±)-4-[4, 5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxy-phenyl)-4, 5-dihydro-imidazole-1-carbonyl]-piperazin-2-one
Molecular Formula C30H30Cl2N4O4
Safety and Handling

Store at -20°C