Please login to your online account to display your discounted pricing

Upchurch Scientific™ Nut

$145.63 - $145.63

Specifications

Color Black
Type Nut
For Use With 1/16 in. O.D. Tubing
View More Specs
Products
Catalog Number Mfr. No. Material Price Quantity    

29-008-179

 
upchurch scientific™
UPF195X
PEEK/Stainless Steel Pack of 10 Each for $145.63
Description & Specifications

Specifications

Color Black
Type Nut
For Use With 1/16 in. O.D. Tubing