Please login to your online account to display your discounted pricing

Nur77 Mouse anti-Mouse, PerCP-eFluor 710, Clone: 12.14, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $477.05

Specifications

Antigen Nur77
Clone 12.14
Host Species Mouse
Gene Alias NGFI-B, NR4A1, TR3, NAK1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123874

 
life technologies
LS46596580
25μg Each for $201.55

501128999

 
life technologies
LS46596582
100μg Each for $477.05
Description & Specifications

Specifications

Antigen Nur77
Clone 12.14
Host Species Mouse
Gene Alias NGFI-B, NR4A1, TR3, NAK1
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This 12.14 monoclonal antibody reacts with mouse Nur77 (also known as NR4A1, TR3, NGFI-B, or NAK1), an inducible orphan nuclear receptor. Expressed in thymocytes and T cell lines, Nur77 promotes apoptosis and plays a role in thymocyte negative selection. Additionally, Nur77 has been shown to be critical for steroid biosynthesis in Leydig cells as well as for the effects of dopamine. In addition, Nur77 has been shown to interact with FoxP3 in regulatory T cells. However, our results with this antibody do not correlate with this observation.