Thermo Scientific™ Nunclon™ Sphera™ Microplates
Make your most valuable work your best work. These microplates feature a superior cell culture surface with performance that elevates stem cell and cell culture research.
$491.12 - $2,573.82
Specifications
Certifications/Compliance Free of any animal products; nonpyrogenic, endotoxin tested (0.5 EU/mL); USP biological reactivity class VI
Sterile SAL 10-6, ethylene oxide gas sterilization
View More Specs
Disclaimers:
Shelf Life
  • 4 years
Products ${productFamilyLength}
Catalog Number Mfr. No. Description No. per Pack Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Description No. per Pack Volume (Metric) Price Quantity  
12-566-430 Thermo Scientific™
N174925
1 300μL Case of 8 Each for $491.12
12-566-431 Thermo Scientific™
N174927
1 400μL Case of 8 Each for $505.91
12-566-432 Thermo Scientific™
N174929
5 300μL Case of 50 Each for $2,573.82
Specifications
Description
With the super low cell attachment surface, Thermo Scientific™ Nunclon™ Sphera™ Microplates consistently promote 3D spheroids and organoids culture, aiding you in cancer spheroid formation and stem cell differentiation for disease modeling and drug screening.
Optimized performance for 3D cell culture
  • Allows cells to grow in suspension with virtually no cell attachment
  • Supports many different cell types to generate spheroids
  • Promotes consistent formation of cancer spheroids that simulate the 3D structures of tumor growth
  • Demonstrates superior quality for embryoid body formation of pluripotent stem cells with minimal spontaneous differentiation
  • Proven efficacy in supporting long-term organoid culture
Certifications/Compliance
  • Animal origin-free
  • Nonpyrogenic: Endotoxin tested (0.5 EU/mL)
  • Biologically inert: USP biological reactivity class VI
  • High level of sterility: SAL 10-6, ethylene oxide gas sterilization
  • Shelf life: 4 years from the date of manufacture
Description & Specifications
Specifications
Free of any animal products; nonpyrogenic, endotoxin tested (0.5 EU/mL); USP biological reactivity class VI
SAL 10-6, ethylene oxide gas sterilization