Thermo Scientific™ Nunc™ White 384-Well Optical Bottom Plates
Perform luminescence measurements and other microscopic applications with these white microplates, featuring maximum reflection and minimal autoluminescence.
$675.69 - $867.40
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Color White
Material Polystyrene
No. per Pack 10
Type Optical Bottom Plate
View More Specs
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Lid Sterile Surface Coating Price Quantity  
Catalog Number Mfr. No. Color Lid Sterile Surface Coating Price Quantity  
12-566-1 Thermo Scientific™
N142762
White Yes Yes Cell Culture Case of 30 Each for $867.40
12-565-355 Thermo Scientific™
242763
White No No Non-Treated Case of 30 Each for $675.69
Specifications
Description
Perform luminescence measurements and other microscopic applications with Thermo Scientific™ Nunc™ White 384-Well Optical Bottom Plates. These microplates have maximum reflection and minimal autoluminescence.
Description & Specifications
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Polystyrene
Optical Bottom Plate
White
10
384 F