Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ MicroWell™ Plates with Nunclon™ Delta Surface

Perform a variety of applications including cloning experiments and tissue culture-based analysis with Nunc MicroWell Plates with Nunclon Delta Surface.

$320.22 - $457.96

Specifications

Certifications/Compliance Each case of plates contains a certificate that guarantees biological performance and traceability.
Culture Area 0.36cm²
Sterile Yes
Well Design 96 U
Well Design 96 U
View More Specs

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Products
Catalog Number Mfr. No. Lid Price Quantity    

12-565-63

 
thermo scientific™
N143761
No Case of 50 Each for $320.22

12-565-65

 
thermo scientific™
N163320
Yes Case of 50 Each for $457.96
Description & Specifications

Specifications

Certifications/Compliance Each case of plates contains a certificate that guarantees biological performance and traceability.
Culture Area 0.36cm²
Sterile Yes
Well Design 96 U
Well Design 96 U
Length (Metric) Exterior 128mm
No. per Case 50
No. per Pack 1
No. of Wells 96
Width (Metric) Exterior 86mm

Perform a variety of applications including cloning experiments and tissue culture-based analysis with Thermo Scientific™ Nunc™ MicroWell™ Plates with Nunclon™ Delta Surface.

Features:

  • Tight-fitting lids reduce evaporation
  • Stackable
  • Radiation sterilized
  • Condensation rings control evaporation and retain condensate
  • Keyed lids prevent accidental reversals

Ordering Information:

Individually wrapped.