Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Polymer Base
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with these 96-well optical-bottom plates with polymer base.
$425.58 - $666.64
Specifications
Material Polymer
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Well Design 96 F
Volume (Metric) Well 400µL
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Color Lid Sterile Surface Coating Price Quantity    
12-565-903
thermo scientific™
N152028
White Yes No Poly-D-Lysine Case of 20 Each for $666.64
12-565-909
thermo scientific™
N152036
Black Yes No Collagen I Case of 20 Each for $609.40
12-565-902
thermo scientific™
N152037
Black Yes No Poly-D-Lysine Case of 20 Each for $587.87
12-566-70
thermo scientific™
N165305
Black Yes Yes Cell Culture Case of 30 Each for $425.58
12-566-71
thermo scientific™
N165306
White Yes Yes Cell Culture Case of 30 Each for $425.58
12-566-72
thermo scientific™
265301
Black No No Non-Treated Case of 30 Each for $454.52
12-566-73
thermo scientific™
N265302
White No No Non-Treated Case of 30 Each for $504.12
12-565-910
thermo scientific™
N152040
White Yes No Collagen I Case of 20 Each for $649.13
Description & Specifications
Specifications
Material Polymer
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Well Design 96 F
Volume (Metric) Well 400µL
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Polymer Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Customized bar coding is available
Ordering Information:
Part No. 152040, 152036, 152028 and 152037 is produced in a clean environment.
Warranty: 90 days