Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with these plates with coverglass base.
$2,842.94 - $3,355.96
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
No. per Case 30
Well Design 96 F
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Color Surface Coating Price Quantity    
12-565-607
thermo scientific™
160376
Black CC2 Case of 30 Each for $3,355.96
12-566-35
thermo scientific™
N164588
Black Cell Culture Case of 30 Each for $3,083.39
12-566-37
thermo scientific™
N164590
White Cell Culture Case of 30 Each for $3,198.27
12-566-38
thermo scientific™
N265300
Black Non-Treated Case of 30 Each for $2,842.94
Description & Specifications
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
No. per Case 30
Well Design 96 F
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Background, autoflorescence and crosstalk are improved
  • Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
  • White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
  • Cell culture surface is ideal for cell-based assays
  • CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
  • Customized bar coding available