Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ High Density Cell Factory™ Systems


Designed to increase cell culture yield and improve productivity in the same footprint as a standard Thermo Scientific™ Nunc™ Cell Factory™ system.

Manufacturer: thermo scientific™  N169102

 View more versions of this product

Catalog No. 12-565-750

  • / Case
 

Description & Specifications

Specifications

Sterile Yes
Description Nunc High Density Cell Factory System
Closure Type Threaded vent port closure
No. per Pack 1
Culture Area 32864cm²
Surface Treatment Nunclon™ Delta
Tray Layers 52
Depth (Metric) 698mm
Dimensions (L x W) 13.2 x 8.1 in. (335 x 205mm)
Material Polystyrene
No. per Case 1
Depth (English) 27.5 in.

Thermo Scientific™ Nunc™ High Density Cell Factory™ System offers 30% more surface area and yield* than the standard Cell Factory system or similar multi-tray systems for adherent cell culture. Its economic benefits are immediate with impacts across many functions of an organization, as it enables end users to optimize their cell culture manufacturing footprint, reduce material usage and labor-consumption as well as to increase their cell culture yield.