Thermo Scientific™ Nunc™ High Density Cell Factory™ Systems
Designed to increase cell culture yield and improve productivity in the same footprint as a standard Thermo Scientific™ Nunc™ Cell Factory™ system.
Manufacturer:  Thermo Scientific™ N169118
 View more versions of this product
Catalog No. 12-565-751
Specifications
Description
Thermo Scientific™ Nunc™ High Density Cell Factory™ System offers 30% more surface area and yield* than the standard Cell Factory system or similar multi-tray systems for adherent cell culture. Its economic benefits are immediate with impacts across many functions of an organization, as it enables end users to optimize their cell culture manufacturing footprint, reduce material usage and labor-consumption as well as to increase their cell culture yield.
Description & Specifications
Specifications
Nunc High Density Cell Factory System
Polystyrene
13
3
7.3 in.
13.2 x 8.1 in. (335 x 205mm)
8216cm²
Yes
1
186mm
Nunclon™ Delta
Threaded vent port closure