Thermo Scientific™ Nunc™ Edge 2.0 96-Well Plates
Reduce edge effect and minimize plate evaporation using this uniquely designed moat plate simply by filling the surrounding reservoir with media or sterile water.
$150.40 - $690.62
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Color Clear
Well Design 96F
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Lid Sterile Surface Coating Price Quantity  
Catalog Number Mfr. No. Lid Sterile Surface Coating Price Quantity  
14387220 Thermo Scientific™
167425
Yes Yes Nunclon Delta Case for $229.44
14387221 Thermo Scientific™
167542
Yes Yes Nunclon Delta Case for $690.62
14387222 Thermo Scientific™
167574
No Yes Nunclon Delta Case for $214.22
14387223 Thermo Scientific™
167554
No Yes Nunclon Delta Case for $577.04
14387224 Thermo Scientific™
267427
Yes Yes Non-Treated Case for $195.82
14387225 Thermo Scientific™
267544
Yes Yes Non-Treated Case for $563.40
14387227 Thermo Scientific™
267556
No Yes Non-Treated Case for $513.42
14387226 Thermo Scientific™
267576
No Yes Non-Treated Case for $150.40
14387228 Thermo Scientific™
267578
Yes No Non-Treated Case for $477.09
14-387-229 Thermo Scientific™
267566
No No Non-Treated Case for $495.24
Specifications
Description
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Reclaim full-use of your 96-well plate
  • The Edge 2.0 Plate provides an ideal, efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
  • Uniquely designed with a surrounding moat that serves as an evaporation barrier, enabling researchers to expand their microplate cultures to all 96 wells without concerns of evaporation.
Achieve full-plate consistency and enhanced productivity
  • Following incubation periods of up to four days, Edge 2.0 yields consistent, viable cells across the entire 96-well plate, leading to improved productivity for cell- based assays.
Improve your process and results
  • Enabling the use of all 96 wells permits experiment efficiencies that can result in potentially less cost and related product waste.
Optimized for extended microplate cultures
  • Nunclon™ Delta cell culture surface promotes cell attachment and growth; untreated surface options available
  • Filling perimeter reservoir with 1.7 sterile water or media helps to ensure well-to-well consistency and cell viability
  • Overall plate evaporation rate reduced to <2% after four days of incubation
  • Total well volume = 400 µL
  • Using the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader, the filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Warranty: 90 days
Description & Specifications
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Clear
96F