missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ Edge 96-Well Plates

Minimize plate evaporation through the uniquely engineered design of Thermo Scientific™ Nunc™ Edge 96 Well Plate

Manufacturer:  Thermo Scientific™ 167314

 View more versions of this product

Catalog No. 12565026


 

Specifications

Description

Minimize plate evaporation through the uniquely engineered design of Thermo Scientific™ Nunc Edge 96-Well Plate. The Edge plate is engineered for optical superiority allowing your automated imaging systems to operate efficiently. Using the Edge Plate in conjunction with Tecan's™ innovative Gas Control module for the Infinite™ 200 PRO multimode reader platform, the filled moats of the plate function as passive humidity control, which allows for simultaneous incubation and signal detection of cells during long-term applications with living cells inside Tecan's multimode reader.

  • Clear Polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment.
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Customized bar coding is available

Reduced Edge Effect

  • Virtually eliminates evaporation in lengthy incubations
  • Use all 96 wells with fewer concerns of edge effects

Low Evaporation Permits Full-Plate Results

  • Unique design minimizes plate evaporation
  • Large evaporative buffer zones eliminate well-to-well variability
  • Overall plate evaporation rate reduced to <2% after seven days of incubation
  • Perimeter moat can be filled with 1.7mL sterile H2O or media
  • Total well volume: 400μL

Optical Properties are Critical

  • Clear plates are engineered for optical superiority
  • Low background fluorescence
  • Extreme well flatness
  • Ideal for image-based high content applications like measuring primary neuron cultures
Description & Specifications

Specifications

Yes
Clear
50
Yes
96 F
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
96
50
Nunclon Delta