Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ Edge 96-Well Plates


Minimize plate evaporation through the uniquely engineered design of Thermo Scientific™ Nunc™ Edge 96 Well Plate

Manufacturer: thermo scientific™  267312

 View more versions of this product

Catalog No. 12565027

  • / Case of 160 Each
 

Description & Specifications

Specifications

Lid Yes
No. of Wells 96
No. per Case 160
No. per Pack 10
Sterile No
Surface Coating Non-Treated
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Well Design 96 F
Color Clear
Lid Yes
Wells 96
No. per Pack 10

  • Clear Polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment.
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Customized bar coding is available

Reduced Edge Effect

  • Virtually eliminates evaporation in lengthy incubations
  • Use all 96 wells with fewer concerns of edge effects

Low Evaporation Permits Full-Plate Results

  • Unique design minimizes plate evaporation
  • Large evaporative buffer zones eliminate well-to-well variability
  • Overall plate evaporation rate reduced to <2% after seven days of incubation
  • Perimeter moat can be filled with 1.7mL sterile H2O or media
  • Total well volume: 400μL

Optical Properties are Critical

  • Clear plates are engineered for optical superiority
  • Low background fluorescence
  • Extreme well flatness
  • Ideal for image-based high content applications like measuring primary neuron cultures