Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated EasYFlasks™
Enhance cell attachment to culture surface with the extracellular matrix coating of Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated EasYFlasks™.
Supplier:  Thermo Scientific™ 132707
 View more versions of this product
Catalog No. 01-814-58
Description
Description
Enhance cell attachment to culture surface with the extracellular matrix coating of Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated EasYFlasks™, ideal for culturing finicky cells that do not attach to the regular cell culture-treated surfaces.
Features
  • Wide opening and angled neck for easy and full access to the culture area
  • Ergonomic closure, open or close in 1/3 turn
  • Ideal for culturing finicky cells that do not bind to regular cell culture treated surface
  • Sterile and non-pyrogenic
Filter Caps:
  • Vent/close and filter caps are available
  • Filter caps are fitted with a hydrophobic filter to ensure consistent gas exchange
  • "Y"-mark allows visual verification of vent position, even when stacked in incubators
Ordering Information:
Extra caps in every case
Includes:
Filter Caps
Recommended for:
Cell culture
Specifications
Specifications
Nunc EasYFlask 75cm2
Angled
5
Wide
Flat
75 cm2
Polystyrene
30
Collagen I, rat tail
Aseptically Manufactured
Filter
25 mL
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.