Thermo Scientific™ Nunc™ Center Well Dish for IVF
Consistently and reliably obtain high-quality results during embryo and oocyte washing with Thermo Scientific™ Nunc™ Center Well Dishes for IVF.
$515.91 - $515.91
Specifications
Description Center Well Dish
Item Description Center well
No. per Case 120
No. per Pack 120
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
12-565-024 Thermo Scientific™
N150260
Center Well Dish Case of 120 Each for $515.91
Specifications
Description
Consistently and reliably obtain high-quality results during embryo and oocyte washing with Thermo Scientific™ Nunc™ Center Well Dishes for IVF.
Innovative features for quality and ease-of-use:
  • Perfect angle of reservoir well walls for locating and easy washing of samples
  • True flat bottom for maintaining optimal temperature
  • Easy grip design for increased safety
Rigorously tested, certified and validated for IVF:
  • CE-marked, FDA-cleared and MEA-tested
  • Produced under ISO 13485 with full lot traceability
Certificates of conformity for your assurance on every lot:
  • Sterile (SAL 10-6)
  • Non-pyrogenic (LAL test)
  • USP Class VI test
  • Release test results (MEA)
Warranty: 90 days
Description & Specifications
Specifications
Center Well Dish
Center well
120
120