Thermo Scientific Nunc™ Cell Factory™ Systems with Active Gassing
Create an ideal environment for culturing oxygen-consuming and pH sensitive cells, with a controlled, contamination-free atmosphere in each culture tray.
$3,690.89 - $3,690.89
Specifications
Description Cell Factory with Active Gassing
Sterile Yes
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Culture Area Tray Layers Price Quantity  
Catalog Number Mfr. No. Culture Area Tray Layers Price Quantity  
12-565-107 Thermo Fisher Scientific
N173238
6320cm² 10 Case of 6 Each for $3,690.89
Specifications
Description
Create an ideal environment for culturing oxygen-consuming and pH sensitive cells using Thermo Scientific™ Nunc™ Cell Factory™ Systems with Active Gassing. Flow system helps ensure a controlled, contamination-free atmosphere in each culture tray by equally distributing the user-specified gas mixture. Available in 4-, 10- and 40-tray models.
  • Achieve greater yields with controlled cell culture environments equally distributed within each layer
  • Optimize production capacity without changing cell culture platforms
  • Develop industrial scale solutions with the utilization of an active gassing Cell Factory™ Incubator
Description & Specifications
Specifications
Cell Factory with Active Gassing
Yes