Learn More
Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps.
$457.23 - $819.69
Catalog Number | Mfr. No. | Culture Area | Neck Style | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Culture Area | Neck Style | Price | Quantity | ||||
12-565-540
|
Thermo Scientific™
178983 |
175 cm2 | Straight |
Case of 32 Each for $720.83
|
|
12-565-57
|
Thermo Scientific™
136196 |
25 cm2 | Angled |
Case of 160 Each for $819.69
|
|
12-565-30
|
Thermo Scientific™
178883 |
175 cm2 | Straight |
Case of 32 Each for $547.83
|
|
12-565-32
|
Thermo Scientific™
178905 |
80 cm2 | Straight |
Case of 50 Each for $457.23
|
|
Description
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth. The filter caps allow constant airflow and minimize chances of contamination.
Features:- Nunclon Delta Certification including testing on four different cell types is provided in each case.
- Short wide necks, straight side walls and 1/3-turn caps are optimized for robotic operation.
- Filter caps ensure constant gas exchange.
- Barcoded 175cm2 flasks with large Code 128 are compatible with automation.
- Sterilized by gamma irradiation and certified to be non-pyrogenic.
Specifications
Filter |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.