Please login to your online account to display your discounted pricing

Nunc™ Edge 96-Well Plates

Minimize plate evaporation through the uniquely engineered design of Thermo Scientific™ Nunc™ Edge 96 Well Plate

$452.69 - $2,415.60

Products
Catalog Number Mfr. No. Price Quantity    

12565025

 
thermo scientific™
167311
Case of 160 Each for $2,415.60

12565026

 
thermo scientific™
167314
Case of 50 Each for $656.18

12565027

 
thermo scientific™
267312
Case of 160 Each for $1,233.04

12565028

 
thermo scientific™
267313
Case of 50 Each for $452.69
Description & Specifications

  • Clear polystyrene has excellent optical properties making it ideal for optical measurements 
  • Untreated surface is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Surfaces are also ideal for adding coatings in your lab
  • Thermo Scientific™ Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Customized bar coding is available
  • Virtually eliminates evaporation in lengthy incubations
  • Use all 96 wells with fewer concerns of edge effects 
 
Low Evaporation Permits Full Plate Results
 
  • Large evaporative buffer zones eliminate well-to-well variability 
  • Overall plate evaporation rate reduced to <2% after seven days of incubation
  • Perimeter moat can be filled with 1.7mL sterile H O or media
  • Total well volume: 400μL
 
Optical Properties are Critical
  • Clear plates are engineered for optical superiority 
  • Low background fluorescence
  • Extreme well flatness
  • Ideal for image¬-based high content applications like measuring