Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ 384-Well Polystyrene Black and White Microplates
Eliminate wicking thanks to the rounded square geometry of black and white 384-well plates, optimized for robotics, plate readers and liquid handlers.
$392.02 - $1,312.61
Specifications
Material Polystyrene
Type 384-Well Microplate
Volume (Metric) Total 120µL
Well Design 384 F
Well Design 384 F
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Color Sterile Surface Coating Price Quantity    
12-565-343
thermo scientific™
164610
White Yes Cell Culture Case of 30 Each for $392.02
12-568-52
thermo scientific™
N165195
White Yes Cell Culture Case of 100 Each for $1,172.13
1256854
thermo scientific™
262260
Black No Non-Treated Case of 100 Each for $969.71
1256855
thermo scientific™
262360
White No Non-Treated Case of 100 Each for $1,312.61
12-565-344
thermo scientific™
N164564
Black Yes Cell Culture Case of 30 Each for $407.24
Description & Specifications
Specifications
Material Polystyrene
Type 384-Well Microplate
Volume (Metric) Total 120µL
Well Design 384 F
Well Design 384 F
Improve background, autofluorescence and crosstalk with Thermo Scientific™ Nunc™ 384-Well Polystyrene Black and White Microplates.
  • Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
Warranty: 90 days