Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ 384-Well Optical Bottom Plates
View microscopic plate contents using luminescence and fluorescence with these 384-well optical bottom plates.
$778.02 - $3,642.29
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Color Black
Volume (Metric) Total 120µL
Well Design 384 F
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Lid No. per Pack Surface Coating Price Quantity    
12-566
thermo scientific™
N142761
Yes 10 Cell Culture Case of 30 Each for $849.07
12-565-905
thermo scientific™
N152029
Yes 5 Poly-D-Lysine Case of 20 Each for $1,292.13
12-565-912
thermo scientific™
N152041
Yes 5 Collagen I Case of 20 Each for $1,195.73
12566280
thermo scientific™
164586**
No 6 Cell Culture Treated Case of 30 Each for $3,642.29
12-565-473
thermo scientific™
164730
No 10 Cell Culture Case of 30 Each for $778.02
12-566-281
thermo scientific™
N240074
No 5 Non-Treated Case of 30 Each for $2,392.13
12-566-2
thermo scientific™
242764
No 10 Non-Treated Case of 30 Each for $925.30
Description & Specifications
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Color Black
Volume (Metric) Total 120µL
Well Design 384 F
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Thermo Scientific™ Nunc™ MicroWell™ 384-Well Optical-Bottom Plates with Polymer or Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Poly- D-Lysine and Collagen coating creates a positive charge on the surface that ensures cell attachment, growth and differentiation.
  • Cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Untreated Polystyrene is hydrophobic and suited for growth of suspension cultures that can proliferate and grow without attachment
  • Polystyrene upper structureis available with either a #1.5 cover glass or polystyrene base
  • Customized bar coding is available
Warranty: 90 days