Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces
Reduce the volume of reagent needed with Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces.
$310.89 - $990.80
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Color Clear
Volume (Metric) Total 120µL
Well Design 384 F,384 F
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
12-565-342 Thermo Scientific™
N164688
Treated; w/lid; Sterile; 10/30 Case of 30 Each for $417.56
12-565-294 Thermo Scientific™
N242757
Non-Treated; w/ lid; Sterile; 10/30 Case of 30 Each for $310.89
12-565-295 Thermo Scientific™
242765
Non-Treated; with lid; Sterile; 1/30 Case of 30 Each for $410.16
12-568-53 Thermo Scientific™
N262160
Non-Treated; no lid; Nonsterile; 25/100 Case of 100 Each for $955.69
12-567-150 Thermo Scientific™
265202
Non-Treated; w/ lid; Sterile; 25/100 Case of 100 Each for $917.51
12-567-151 Thermo Scientific™
265203
Non-Treated; no lid; Sterile; 25/100 Case of 100 Each for $990.80
Specifications
Description
Reduce the volume of reagent needed with Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces, which are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample volume reduction, providing cost savings and allowing more samples to be screened.
Features:
  • Clear Polystyrene has excellent optical properties, making it ideal for optical measurements
  • Untreated Polystyrene is hydrophobic and suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces ideal for adding coatings in the lab
  • Cell culture treated, hydrophilic surface promotes cell attachment and growth
  • Customized bar coding is available
Warranty: 90 days
Description & Specifications
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Clear
120µL
384 F,384 F