Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ 1536 Well High Base Microplates
Work with small volumes and screen 4X as many samples in one plate. Thermo Scientific™ Nunc™ 1536 Well High Base Microplates are ideal for high-throughput screening.
$2,575.16 - $4,336.09
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Well Design 1536 F
No. per Case 100
No. per Pack 25
Volume (Metric) Total 13.4µL
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Color Surface Coating Price Quantity    
12-565-499
thermo scientific™
N164708
Black Cell Culture Case of 100 Each for $2,575.16
12-565-520
thermo scientific™
N264711
Black Non-Treated Case of 100 Each for $2,947.76
12-565-490
thermo scientific™
N264712
White Non-Treated Case of 100 Each for $4,336.09
03-150-12
thermo scientific™
164709
White Cell Culture Case of 100 Each for $2,582.62
Description & Specifications
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Well Design 1536 F
No. per Case 100
No. per Pack 25
Volume (Metric) Total 13.4µL
Work with small volumes and screen 4X as many samples in one plate. Thermo Scientific™ Nunc™ 1536 Well High Base Microplates are ideal for high-throughput screening; the skirted design around the base of the plate is optimal for robotic handling.
 Warranty: 90 days