Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ 1.8mL Externally-Threaded Universal Tubes
Use this versatile tube either inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars.
Manufacturer: thermo scientific™  N331830
 View more versions of this product
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Catalog No. 12-590-009
  • / Case of 10 Each
Description & Specifications
Specifications
Color Beige with blue latches
Description Nunc latch rack
Includes ----
Barcode No
Sterile Sterile
Packaging 10 empty racks
Code Type ----
No. per Pack 1
Capacity (Metric) ----
Vial Type ----
Closure Color ----
No. per Case 10
Quantity 10
Use versatile Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes either inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes, non-coded or both 1D (linear) plus 2D barcodes to serve every need.
Scalable
  • Tubes and rack are ideal for both lower-throughput  and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cyrostorage boxes made for 10x10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack
Innovative Universal Cap Design
  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
  • Permanently-bonded,unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
Stringent Quality Control
  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 10superscript 6 and is made with class VI medical grade resins