Please login to your online account to display your discounted pricing

Alfa Aesar™ Nullscript, 97%


Quantity 1mg

Manufacturer: alfa aesar™  J67363MCR

 View more versions of this product

Catalog No. AAJ67363MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point 171° to 172°C
Formula Weight 298.3
Chemical Name or Material Nullscript
CAS 300816-11-9
Assay 97%
MDL Number MFCD00405081
Storage Note 1 -30° to -10°C
Physical Form Solid
Synonym 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide
Color White
Sensitivity Light Sensitive
Quantity 1mg

Specifications

Chemical Name or Material Nullscript
CAS 300816-11-9
Formula Weight 298.3
MDL Number MFCD00405081
Synonym 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide
Accessories

-30° to -10°C