Please login to your online account to display your discounted pricing

Alfa Aesar™ Nullscript, 97%

Chemical Identifiers

Chemical Name or Material Nullscript
CAS 300816-11-9
Formula Weight 298.3
MDL Number MFCD00405081
Synonym 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67363MCR

 
alfa aesar™
J67363MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 298.3
Melting Point 171° to 172°C
Chemical Name or Material Nullscript
Assay 97%
MDL Number MFCD00405081
CAS 300816-11-9
Physical Form Solid
Synonym 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide
Storage Note 1 -30° to -10°C
Color White
Sensitivity Light Sensitive
Quantity 1mg
Safety and Handling

-30° to -10°C