Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Nucleophosmin (NPM) Ab-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Nucleophosmin (NPM) Ab-1, Mouse Monoclonal Antibody.

$248.12 - $1,166.08

Specifications

Antigen Nucleophosmin (NPM) Ab-1
Clone NA24
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1849P

 
thermo scientific
MS1849P
Purified Ab with BSA and Azide 1mL Each for $1,078.12

MS1849P0

 
thermo scientific™ lab vision™
MS1849P0
Purified Ab with BSA and Azide 100µL Each for $248.12

MS1849P1

 
thermo scientific™ lab vision™
MS1849P1
Purified Ab with BSA and Azide 500µL Each for $713.63

MS1849P1ABX

 
thermo scientific™ lab vision™
MS1849P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $784.84

MS1849PABX

 
thermo scientific™ lab vision™
MS1849PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,166.08

MS1849R7

 
thermo scientific™ lab vision™
MS1849R7
Ready-to-use for IHC (UVLP) 7mL Each for $478.06
Description & Specifications

Specifications

Antigen Nucleophosmin (NPM) Ab-1
Clone NA24
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen GST fusion protein containing the N-terminal part of nucleophosmin fused to 14 aa of ALK protein
Isotype IgG kappa

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Nucleophosmin (NPM) Ab-1, Mouse Monoclonal Antibody.

Nucleophosmin (NPM)/B23/numatrin is a phosphoprotein associated with cell nucleoli. Treatment of cells with cytotoxic drugs may make nucleophosmin enter nucleoplasm. It is reported that nucleophosmin shuttles continuously between cytoplasm and nucleoli to carry newly synthesized ribosomal proteins into nucleoli. NPM-ALK is a hybrid of the anaplastic lymphoma kinase (ALK) gene and the nucleophosmin (NPM) gene resulting from the t(2;5)(p23;q35) translocation found in anaplastic large cell lymphoma. In acute promyelocytic leukemia NPM gene is fused to RARalpha gene in (5;17) (q33;q12) transition.

Host Species: Mouse

Clone: NA24

Isotype: IgG/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: GST fusion protein containing the N-terminal part of nucleophosmin fused to 14aa of ALK protein

Molecular Weight: 38kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO