missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nuclear Fast Red [for Determination of Ca in Serum], TCI America™

$68.75 - $204.72

Chemical Identifiers

CAS 6409-77-4
Molecular Formula C14H11NNaO7S
Molecular Weight (g/mol) 360.292
MDL Number MFCD00078493
InChI Key KDEATZKCHKVXGA-UHFFFAOYSA-N
Synonym Kernechtrot, NFR
PubChem CID 131675027
IUPAC Name 4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium
SMILES [HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0305100MG View Documents TCI America
N0305100MG
100mg Each for $68.75
 
N03051G View Documents TCI America
N03051G
1g Each for $204.72
 
Specifications

Chemical Identifiers

6409-77-4
360.292
KDEATZKCHKVXGA-UHFFFAOYSA-N
131675027
[HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]
C14H11NNaO7S
MFCD00078493
Kernechtrot, NFR
4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium

Specifications

6409-77-4
MFCD00078493
KDEATZKCHKVXGA-UHFFFAOYSA-N
4-amino-1,3-dihydroxy-9,10-dioxoanthracene-2-sulfonic acid;molecular hydrogen;sodium
131675027
Crystal-Powder at 20°C
100mg
C14H11NNaO7S
Kernechtrot, NFR
[HH].C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C(=C3N)O)S(=O)(=O)O)O.[Na]
360.292
357.27
Undesignated
Nuclear Fast Red [for Determination of Ca in Serum]
Safety and Handling

Safety and Handling

TSCA : Yes