Invitrogen™ NUAK1 (ARK5) Recombinant Human Protein DFS Item
NUAK1 (ARK5) is a serine/threonine kinase that plays a role in many cellular functions.
Manufacturer:  Invitrogen™ LSPV4283
 View more versions of this product
Catalog No. PV4283
Description
Description
The overexpression of the NUAK1 (ARK5) kinase has been associated with tumor invasion and metastasis (colorectal neoplasm).
Specifications
Specifications
NP_055655
Frozen
78.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
CAMKL Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
0mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% TritonX-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
NUAK1 (ARK5)
Baculovirus
Human
His Tag
Documents