Please login to your online account to display your discounted pricing

NU 6140, Tocris

Cyclin-dependent kinase 2 (cdk2) inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material NU 6140
Assay Percent Range >99%
Molecular Formula C23H30N6O2
Formula Weight 422.52
Synonym 4-[[6-(Cyclohexylmethoxy)-1H-purin-2-yl]amino]-N, N-diethylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

330110

 
r & d systems
330110
10mg Each for $252.39

330150

 
r & d systems
330150
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 422.52
Quantity 10mg
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C
Synonym 4-[[6-(Cyclohexylmethoxy)-1H-purin-2-yl]amino]-N, N-diethylbenzamide
Molecular Formula C23H30N6O2
Chemical Name or Material NU 6140
CAS 444723-13-1
Safety and Handling

Desiccate at +4°C