Please login to your online account to display your discounted pricing

NU 2058, Tocris

Cdk1 and cdk2 inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material NU 2058
Assay Percent Range >99%
Molecular Formula C12H17N5O
Formula Weight 247.3
Synonym 6-(Cyclohexylmethoxy)-9H-purin-2-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

313510

 
r & d systems
313510
10mg Each for $201.36

313550

 
r & d systems
313550
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 247.3
Quantity 10mg
Chemical Name or Material NU 2058
Assay Percent Range >99%
CAS 161058-83-9
Molecular Formula C12H17N5O
Synonym 6-(Cyclohexylmethoxy)-9H-purin-2-amine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C